Tetracycline

Supplier: Thermo Scientific Chemicals
Warning

J61714-06 J61714-14 J61714-22
AAJ61714-06EA 49.37 USD
AAJ61714-06 AAJ61714-14 AAJ61714-22
Tetracycline
Tetracycline

Can induce apoptosis in osteoclasts


Crystalline powder


Can induce apoptosis in osteoclasts

Formula: C₂₂H₂₄N₂O₈
MW: 444.43 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00151232
CAS Number: 60-54-8
EINECS: 200-481-9
Merck Index: 14,9196

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR