Dobutamine hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
A beta-1 adrenoceptor agonist with weak beta-2 adrenoceptor activity and some action at the alpha-1 adrenoceptor
Off-white solid
Applications: A beta-1 adrenoceptor agonist with weak beta-2 adrenoceptor activity and some action at the alpha-1 adrenoceptor
Formula:
C₁₈H₂₃NO₃ Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153795 CAS Number: 49745-95-1 EINECS: 256-464-1 Merck Index: 14,3395 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...