(+)-1-Deoxynojirimycin hydrochloride
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Deoxymannojirimycin hydrochloride
J61277-LB0
J61277-MA
AAJ61277-LB0EA
175
USD
AAJ61277-LB0
AAJ61277-MA
AAJ61277-MB
(+)-1-Deoxynojirimycin hydrochloride
(+)-1-Deoxynojirimycin hydrochloride
Selective inhibitor of a-mannosidase
CAS No.: 73465-43-7
Molecular Formula: C6H13NO4.HCl
Formula Weight: 199.60
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00083611
Selective inhibitor of a-mannosidase
Formula:
C₆H₁₄ClNO₄ MW: 199.64 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00083611 CAS Number: 73465-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...