Paromomycin sulfate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate(1:x)
J61274-03
J61274-06
J61274-14
AAJ61274-03EA
55.97
USD
AAJ61274-03
AAJ61274-06
AAJ61274-14
Paromomycin sulfate
Paromomycin sulfate
Inhibits initiation and elongation during protein synthesis
White to off-white powder
Applications: Inhibits initiation and elongation during protein synthesis
Formula:
C₂₃H₄₅N₅O₁₄·H₂SO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00079278 CAS Number: 1263-89-4 EINECS: 215-031-7 Merck Index: 14,7041 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...