Kanamycin sulfate

Supplier: Thermo Scientific Chemicals
Danger

J61272-09 J61272-14 J61272-22
AAJ61272-09EA 210.68 USD
AAJ61272-09 AAJ61272-14 AAJ61272-22
Kanamycin sulfate
Kanamycin sulphate

Antibiotic. Inhibits translocation during protein biosynthesis


Powder


Antibiotic. Inhibits translocation during protein biosynthesis

Formula: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9
Merck Index: 14,5281

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR