2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Amfebutamone hydrochloride, Bupropion Hydrochloride, Bupropion hydrochloride, 2-(tert-Butylamino)-1-(3-chlorophenyl)-1-propanone hydrochloride

J61105-03 J61105-MD
AAJ61105-03EA 175 USD
AAJ61105-03 10814-244
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride 99%
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride

A selective inhibitor of dopamine uptake


Powder


Applications: A selective inhibitor of dopamine uptake

Formula: C13H19Cl2NO
MW: 276.20
Melting Pt: 233…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00055209
CAS Number: 31677-93-7
EINECS: 250-759-9
Merck Index: 14,1499

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR