Tamoxifen citrate

Supplier: Thermo Scientific Chemicals
Danger

J60955-03 J60955-06
AAJ60955-03EA 202 USD
AAJ60955-03 AAJ60955-06
Tamoxifen citrate
Tamoxifen citrate

Inhibitor of protein kinase C and induces apoptosis in cancer cells


White to off-white powder


Applications: Inhibitor of protein kinase C and induces apoptosis in cancer cells

Formula: C₂₆H₂₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD00058321
CAS Number: 54965-24-1
EINECS: 259-415-2
Merck Index: 14,9048

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR