SB202190 ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J60950-MB J60950-MF
AAJ60950-MBEA 141 USD
AAJ60950-MB AAJ60950-MC AAJ60950-MF
SB202190 ≥99%
SB202190

Inhibits p38 kinase activity in vivo through competition with ATP


Powder

Formula: C₂₀H₁₄FN₃O
Melting Pt: 240…243 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941964
CAS Number: 152121-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR