Sodium phosphate, 0.5 M buffer soln., pH 8.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60825-AK
J60825-AP
AAJ60825-AKEA
208.97
USD
AAJ60825-AK
AAJ60825-AP
Sodium phosphate, 0.5 M buffer soln., pH 8.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...