Hyaluronic acid (from bovine vitreous humor)

Supplier: Thermo Scientific Chemicals

J60566-MA J60566-MC J60566-MF
AAJ60566-MAEA 250.99 USD
AAJ60566-MA AAJ60566-MC AAJ60566-MF
Hyaluronic acid (from bovine vitreous humor)
Hyaluronic acid

A natural high-viscosity polymer of acetylglucosamine and glucuronic acid


Lyophilized powder


Applications: A natural high-viscosity polymer of acetylglucosamine and glucuronic acid

Formula: C₂₈H₄₄N₂O₂₃
Storage Temperature: Ambient
MDL Number: MFCD00131348
CAS Number: 9004-61-9
EINECS: 232-678-0
Merck Index: 14,4757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR