Amiodarone hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Amiodarone HCl, (2-Butylbenzofuran-3-yl)(4-(2-(diethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride)
J60456-03
J60456-06
J60456-14
AAJ60456-03EA
21.04
USD
AAJ60456-03
AAJ60456-06
AAJ60456-14
Amiodarone hydrochloride
Amiodarone hydrochloride
A non-selective ion channel blocker; also an antiarrhythmic agent
Powder
Applications: A non-selective ion channel blocker; also an antiarrhythmic agent
Formula:
C₂₅H₂₉I₂NO₃ Melting Pt: 156…163 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00069204 CAS Number: 19774-82-4 EINECS: 243-293-2 Merck Index: 14,482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...