β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
β-NADPH tetrasodium salt tetrahydrate
Applications: beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.
- Store in cool place
- Keep container tightly closed in a dry and well-ventilated place
- Light sensitive
- Hygroscopic
Purity: 98+%
Molecular Formula: C21H26N7Na4O17P3.xH2O
Formula Weight: 905.42 (833.35anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD07366294
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...