Chenodeoxycholic acid

Supplier: Thermo Scientific Chemicals
Warning

J60364-06 J60364-14
AAJ60364-06EA 90.4 USD
AAJ60364-06 AAJ60364-14
Chenodeoxycholic acid
Chenodeoxycholic acid

Bile acid that induces apoptosis through protein kinase C signaling pathways


Needle crystals


Bile acid that induces apoptosis through protein kinase C signaling pathways

Formula: C₂₄H₄₀O₄
Melting Pt: 164…168 °C
Density: 1.162 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064142
CAS Number: 474-25-9
EINECS: 207-481-8
Merck Index: 12,02096

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR