Chondroitin sulfate ≥90%

Supplier: Thermo Scientific Chemicals

J60341-03 J60341-06 J60341-14
AAJ60341-03EA 48.09 USD
AAJ60341-03 AAJ60341-06 AAJ60341-14
Chondroitin sulfate ≥90%
Chondroitin sulfate

A mucopolysaccharide with N-acetylchondrosine as a repeating unit and with one sulfate group per disaccharide unit; acts as the flexible connecting matrix between the tough protein filaments in cartilage to form a polymeric system.A naturocetic agent


Powder


Applications: A mucopolysaccharide with N-acetylchondrosine as a repeating unit and with one sulfate group per disaccharide unit; acts as the flexible connecting matrix between the tough protein filaments in cartilage to form a polymeric system.A naturocetic agent

Storage Temperature: Ambient
MDL Number: MFCD00146419
CAS Number: 9007-28-7
EINECS: 232-696-9
Merck Index: 14,2214

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR