3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate

J60332-03 J60332-06 J60332-88
AAJ60332-03EA 99.3 USD
AAJ60332-03 AAJ60332-06 AAJ60332-88
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 98%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.


Powder


Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

Formula: C₁₆H₂₀N₂·2HCl·xH₂O
MW: 313.27 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150104
CAS Number: 207738-08-7
EINECS: 264-769-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR