Potassium phosphate, 0.5 M buffer soln., pH 7.2, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60326-AK
J60326-AP
AAJ60326-AKEA
93.6
USD
AAJ60326-AK
AAJ60326-AP
Potassium phosphate, 0.5 M buffer soln., pH 7.2, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...