Tyrphostin A23 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(3,4-Dihydroxybenzylidene)malononitrile

J60308-LB0 J60308-MA J60308-MB
AAJ60308-LB0EA 81.7 USD
AAJ60308-LB0 AAJ60308-MA AAJ60308-MB
Tyrphostin A23 99%
Tyrphostin A23

Potent inhibitor of EGF receptor kinase activity


Powder


Potent inhibitor of EGF receptor kinase activity

Formula: C₁₀H₆N₂O₂
MW: 186.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133899
CAS Number: 118409-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR