SOC medium, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60255-AE
J60255-AK
AAJ60255-AEEA
166.49
USD
AAJ60255-AE
AAJ60255-AK
SOC medium, Thermo Scientific
Cell Culture Media
Liquid
Physical Form: Liquid
MDL No.: MFCD00284637
Applications: For cultivating recombinant strains of Escherichia coli
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...