Sodium fluoride 200 mM in aqueous solution
Supplier: Thermo Scientific Chemicals
J60251-AE
J60251-AK
AAJ60251-AEEA
71.1
USD
AAJ60251-AE
AAJ60251-AK
Sodium fluoride 200 mM in aqueous solution
Sodium fluoride
Liquid
CAS No.: 7681-49-4
Molecular Formula: FNa
Formula Weight: 41.99
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00003524
Formula:
NaF MW: 41.99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...