Canrenone 98%

Supplier: Thermo Scientific Chemicals

J60238-03 J60238-06
AAJ60238-03EA 64.7 USD
AAJ60238-03 AAJ60238-06 AAJ60238-14
Canrenone 98%
Canrenone

Inhibits aldosterone biosynthesis and blocks ouabain effects


Solid


Applications: Inhibits aldosterone biosynthesis and blocks ouabain effects

Formula: C₂₂H₂₈O₃
MW: 340.46 g/mol
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00071735
CAS Number: 976-71-6
EINECS: 213-554-5
Merck Index: 14,1752

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR