Lactulose 99%
Supplier: Thermo Scientific Chemicals
J60160-14
J60160-18
J60160-22
AAJ60160-14EA
330.08
USD
AAJ60160-14
AAJ60160-18
AAJ60160-22
Lactulose 99%
Lactulose
A synthetic sugar
Powder
Applications: A synthetic sugar
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 169 °C Density: 1.32 Storage Temperature: Ambient |
MDL Number:
MFCD00151469 CAS Number: 4618-18-2 EINECS: 225-027-7 Merck Index: 14,5346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...