Sodium phosphate, 0.5 M buffer soln., pH 7.6, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60158-AK J60158-AP
AAJ60158-AKEA 107 USD
AAJ60158-AK AAJ60158-AP
Sodium phosphate, 0.5 M buffer soln., pH 7.6, Thermo Scientific
Buffers Biological Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00146206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR