Psoralen 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7H-Furo[3,2-g]chromen-7-one, Ficusin

J60042-MC J60042-ME
AAJ60042-MCEA 265 USD
AAJ60042-MC AAJ60042-ME
Psoralen 97%
Psoralen

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure


Solid


Applications: Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

Formula: C₁₁H₆O₃
MW: 186.17 g/mol
Melting Pt: 163…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00010520
CAS Number: 66-97-7
EINECS: 200-639-7
Merck Index: 14,7928

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR