Magnesium acetate 1 M in aqueous solution

Supplier: Thermo Scientific Chemicals

J60041-AD J60041-AE
AAJ60041-ADEA 88.32 USD
AAJ60041-AD AAJ60041-AE
Magnesium acetate 1 M in aqueous solution
Magnesium acetate

Liquid


CAS No.: 142-72-3 Molecular Formula: C4H6MgO4 Formula Weight: 142.39 Physical Form: Liquid MDL No.: MFCD00003539

Formula: (H₃CCOO)₂Mg
MW: 142.39
Melting Pt: 323
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00003539
CAS Number: 142-72-3
Merck Index: 12,05688

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR