1-Methylguanidine xhydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methylguanidine hydrochloride

J60033-06 J60033-14
AAJ60033-06EA 84.6 USD
AAJ60033-06 AAJ60033-14
1-Methylguanidine xhydrochloride 98%
1-Methylguanidine xhydrochloride

Crystalline Powder


Purity: 98% CAS No.: 22661-87-6 Molecular Formula: C2H7N3.HCl Formula Weight: 109.56 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Sensitivity: Hygroscopic Appearance: White to off-white MDL No.: MFCD00012576

Formula: C₂H₈ClN₃
MW: 109.557 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012576
CAS Number: 22661-87-6
EINECS: 245-147-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR