Boc-D-Pyr-OH 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid

H58701-03 H58701-06
AAH58701-03EA 47.8 USD
AAH58701-03 AAH58701-06 AAH58701-MD
Boc-D-Pyr-OH 97%
Boc-D-Pyr-OH

Purity: 97% CAS No.: 160347-90-0 Molecular Formula: C10H15NO5 Formula Weight: 229.23 Storage Temperature: 2°C to 8°C Melting Point: 111-116° Optical Rotation: +35° (c=1 in chloroform) MDL No.: MFCD08458561

Formula: C₁₀H₁₅NO₅
MW: 229.232 g/mol
Melting Pt: 111…116 °C
Storage Temperature: Ambient
MDL Number: MFCD08458561
CAS Number: 160347-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR