2-Oxa-7-azaspiro[3.5]nonane hemioxalate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57834-ME
H57834-MD
AAH57834-MEEA
527
USD
AAH57834-ME
AAH57834-MD
2-Oxa-7-azaspiro[3.5]nonane hemioxalate 97%
2-Oxa-7-azaspiro[3.5]nonane hemioxalate
Purity: 97%
CAS No.: 1429056-28-9
Molecular Formula: C7H13NO.0.5C2H2O4
Formula Weight: 172.20
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Density: 1.048
Appearance: White to grey
Melting Point: 208°
MDL No.: MFCD11035888
Formula:
C₁₆H₂₈N₂O₆ Melting Pt: 208 °C Density: 1.048 Storage Temperature: Ambient |
MDL Number:
MFCD11035888 CAS Number: 1429056-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...