4-Bromo-3-fluorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57833-03
H57833-06
AAH57833-03EA
88.6
USD
AAH57833-03
AAH57833-06
AAH57833-MD
4-Bromo-3-fluorobenzaldehyde 97%
4-Bromo-3-fluorobenzaldehyde
Purity: 97%
CAS No.: 133059-43-5
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Melting Point: 54-60°
Beilstein Reference: 9321564
Formula:
C₇H₄BrFO MW: 203.01 g/mol Melting Pt: 54…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095000 CAS Number: 133059-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...