6-Oxa-1-azaspiro[3.3]heptane hemioxalate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Oxa-1-azaspiro[3.3]heptane oxalate (2:1), 6-Oxa-1-azaspiro[3.3]heptane oxalate(2:1)
H57658-ME
H57658-MD
AAH57658-MEEA
714
USD
AAH57658-ME
AAH57658-MD
6-Oxa-1-azaspiro[3.3]heptane hemioxalate 95%
6-Oxa-1-azaspiro[3.3]heptane hemioxalate
Purity: 95%
CAS No.: 1380571-72-1
Molecular Formula: C5H9NO.0.5C2H2O4
Formula Weight: 144.15
Storage Temperature: 2°C to 8°C
MDL No.: MFCD22373634
Formula:
C₁₂H₂₀N₂O₆ MW: 288.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22373634 CAS Number: 1380571-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...