2-Oxa-6-azaspiro[3.4]octane 95%

Supplier: Thermo Scientific Chemicals
Warning

H57579-MD H57579-ME
AAH57579-MDEA 295 USD
AAH57579-MD AAH57579-ME
2-Oxa-6-azaspiro[3.4]octane 95%
2-Oxa-6-azaspiro[3.4]octane

Purity: 95% CAS No.: 220290-68-6 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 63°(145°F) Boiling Point: 187° MDL No.: MFCD14586455

Formula: C₆H₁₁NO
Boiling Pt: 187 °C
Density: 1.08
Flash Pt: 63 °C(145 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD14586455
CAS Number: 220290-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR