Methyl-2-oxoindane-1-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57535-03
H57535-06
AAH57535-03EA
62.6
USD
AAH57535-03
AAH57535-06
Methyl-2-oxoindane-1-carboxylate 97%
Methyl-2-oxoindane-1-carboxylate
Purity: 97%
CAS No.: 104620-34-0
Molecular Formula: C11H10O3
Formula Weight: 190.20
Storage Temperature: 2°C to 8°C
Melting Point: 63-70°
MDL No.: MFCD06200715
Formula:
C₁₁H₁₀O₃ Melting Pt: 63…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200715 CAS Number: 104620-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...