(R)-(-)-Nipecotic acid ≥96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-Piperidine-3-carboxylic acid, (R)-(-)-3-Piperidinecarboxylic acid, D-Nipecotic acid

H57434-14 H57434-03
AAH57434-14EA 759 USD
AAH57434-14 AAH57434-03
(R)-(-)-Nipecotic acid ≥96%
(R)-(-)-Nipecotic acid

Purity: 96+% CAS No.: 25137-00-2 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Melting Point: 250-255° Optical Rotation: -5° (c=1 in water)

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 250…255 °C
Storage Temperature: Ambient
MDL Number: MFCD01630787
CAS Number: 25137-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR