[3-(Bromomethyl)oxetan-3-yl]methanol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57413-MD
H57413-ME
AAH57413-MDEA
53.6
USD
AAH57413-MD
AAH57413-ME
[3-(Bromomethyl)oxetan-3-yl]methanol 95%
[3-(Bromomethyl)oxetan-3-yl]methanol
Purity: 95%
CAS No.: 22633-44-9
Molecular Formula: C5H9BrO2
Formula Weight: 181.03
Flash Point: 100°(212°F)
Boiling Point: 242°
MDL No.: MFCD09800598
Formula:
C₅H₉BrO₂ Boiling Pt: 242 °C Density: 1.598 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD09800598 CAS Number: 22633-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...