Ethyl-2-methylpyridine-4-carboxylate hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Danger

H57239-03 H57239-06
AAH57239-03EA 60.3 USD
AAH57239-03 AAH57239-06 AAH57239-MD
Ethyl-2-methylpyridine-4-carboxylate hydrochloride 95%
Ethyl-2-methylpyridine-4-carboxylate hydrochloride

Purity: 95% CAS No.: 65550-32-5 Molecular Formula: C9H11NO2.HCl Formula Weight: 201.65 Melting Point: 145-151° MDL No.: MFCD09991690

Formula: C₉H₁₁NO₂·HCl
MW: 201.65 g/mol
Melting Pt: 145…151 °C
Storage Temperature: Ambient
MDL Number: MFCD09991690
CAS Number: 65550-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR