2,7-Dibromo-9,9-dihexylfluorene 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,7-Dibromo-9,9-dihexyl-9H-fluorene
H56952-14
H56952-22
AAH56952-14EA
193
USD
AAH56952-14
AAH56952-22
2,7-Dibromo-9,9-dihexylfluorene 98%
2,7-Dibromo-9,9-dihexylfluorene
Purity: 98%
CAS No.: 189367-54-2
Molecular Formula: C25H32Br2
Formula Weight: 492.33
Melting Point: 67-71°
MDL No.: MFCD03427215
Formula:
C₂₅H₃₂Br₂ MW: 492.34 g/mol Melting Pt: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427215 CAS Number: 189367-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...