(4-Nitrophenyl)-β-D-galactopyranoside 98+%

Supplier: Thermo Scientific Chemicals

H56905-06 H56905-09
AAH56905-06EA 604 USD
AAH56905-06 AAH56905-09
(4-Nitrophenyl)-β-D-galactopyranoside 98+%
(4-Nitrophenyl)-β-D-galactopyranoside

Purity: 98+% CAS No.: 3150-24-1 Molecular Formula: C12H15NO8 Formula Weight: 301.25 Storage Temperature: -30°C to -10°C

Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063256
CAS Number: 3150-24-1
EINECS: 221-584-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR