2,4,6-Trichloropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H56787-06
H56787-14
AAH56787-06EA
104
USD
AAH56787-06
AAH56787-14
2,4,6-Trichloropyridine 97%
2,4,6-Trichloropyridine
Purity: 97%
CAS No.: 16063-69-7
Molecular Formula: C5H2Cl3N
Formula Weight: 182.44
Flash Point:
Formula:
C₅H₂Cl₃N MW: 182.44 g/mol Melting Pt: 33…37 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00971928 CAS Number: 16063-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...