2-Phenyl-1,3-propanediol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Phenylpropane-1,3-diol

H56761-06 H56761-14
AAH56761-06EA 114 USD
AAH56761-06 AAH56761-14
2-Phenyl-1,3-propanediol 98%
2-Phenyl-1,3-propanediol

Purity: 98% CAS No.: 1570-95-2 Molecular Formula: C9H12O2 Formula Weight: 152.19 Melting Point: 53-56° MDL No.: MFCD00236056

Formula: HOH₂CCH(C₆H₅)CH₂OH
MW: 152.19 g/mol
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00236056
CAS Number: 1570-95-2
EINECS: 411-810-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR