Bromomethyl acetate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56755-06
H56755-09
AAH56755-06EA
267
USD
AAH56755-06
AAH56755-09
Bromomethyl acetate 95%
Bromomethyl acetate
Purity: 95%
CAS No.: 590-97-6
Molecular Formula: C3H5BrO2
Formula Weight: 152.98
Storage Temperature: 2°C to 8°C
Flash Point: 57°(135°F)
Boiling Point: 130-133°
UN No.: 3272
MDL No.: MFCD00000170
Formula:
H₃CCOOCH₂Br MW: 152.98 g/mol Boiling Pt: 130…133 °C Density: 1.56 Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000170 CAS Number: 590-97-6 EINECS: 209-696-2 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...