4-Chloroquinoline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56721-03
H56721-06
AAH56721-03EA
108
USD
AAH56721-03
AAH56721-06
4-Chloroquinoline 99%
4-Chloroquinoline
Purity: 99%
CAS No.: 611-35-8
Molecular Formula: C9H6ClN
Formula Weight: 163.60
Flash Point: 113°(235°F)
Melting Point: 28-31°
Boiling Point: 260-261°
Beilstein Reference: 114721
MDL No.: MFCD00006773
Formula:
C₉H₆ClN MW: 163.61 g/mol Boiling Pt: 260 - 261 °C (1013 hPa) Melting Pt: 28 °C Density: 1.25 g/cm³ (20 °C) Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006773 CAS Number: 611-35-8 EINECS: 210-267-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...