3,4-Dimethoxythiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

H56674-03 H56674-06
AAH56674-03EA 51.1 USD
AAH56674-03 AAH56674-06
3,4-Dimethoxythiophene 98%
3,4-Dimethoxythiophene

Purity: 98% CAS No.: 51792-34-8 Molecular Formula: C6H8O2S Formula Weight: 144.19 Flash Point: 106°(223°F) Refractive Index: 1.5409 Boiling Point: 100-102°/10-11mm MDL No.: MFCD01096546

Formula: C6H8O2S
MW: 144.19 g/mol
Boiling Pt: 100…102 °C/10…11 mm
Density: 1.209
Flash Pt: 106 °C(223 °F)
Storage Temperature: Ambient
MDL Number: MFCD01096546
CAS Number: 51792-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR