Sodium-4-hydroxybenzoate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Hydroxybenzoate sodium, Sodium 4-hydroxybenzoate, p-Hydroxybenzoic acid Monosodium Salt, 4-Hydroxybenzoic Acid Sodium Salt
H56590-22
H56590-30
AAH56590-22EA
189
USD
AAH56590-22
AAH56590-30
Sodium-4-hydroxybenzoate ≥99%
Sodium-4-hydroxybenzoate
Purity: 99+%
CAS No.: 114-63-6
Molecular Formula: C7H6NaO3
Formula Weight: 160.10
Beilstein Reference: 4163553
MDL No.: MFCD00016530
Formula:
C₇H₆O₃ MW: 160.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016530 CAS Number: 114-63-6 EINECS: 204-051-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...