5,5'-Dibromo-2,2'-bithiophene 99%

Supplier: Thermo Scientific Chemicals
Warning

H56337-06 H56337-14
AAH56337-06EA 86.4 USD
AAH56337-06 AAH56337-14
5,5'-Dibromo-2,2'-bithiophene 99%
5,5'-Dibromo-2,2'-bithiophene

Purity: 99% CAS No.: 4805-22-5 Molecular Formula: C8H4Br2S2 Formula Weight: 324.06 Melting Point: 144-146° MDL No.: MFCD00219110

Formula: C₈H₄Br₂S₂
MW: 324.05 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00219110
CAS Number: 4805-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR