9,9-Di-n-dodecyl-2,7-dibromofluorene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,7-Dibromo-9,9-didodecyl-9H-fluorene

H56320-14 H56320-22
AAH56320-14EA 169 USD
AAH56320-14 AAH56320-22
9,9-Di-n-dodecyl-2,7-dibromofluorene 98%
9,9-Di-n-dodecyl-2,7-dibromofluorene

Purity: 98% CAS No.: 286438-45-7 Molecular Formula: C37H56Br2 Formula Weight: 660.65 Flash Point: 110°(230°F) Melting Point: 50-55° Boiling Point: 180°/0.15mm MDL No.: MFCD03427215

Formula: C₃₇H₅₆Br₂
MW: 660.65 g/mol
Boiling Pt: 180 °C/0.15 mm
Melting Pt: 50…55 °C
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03427215
CAS Number: 286438-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR