DIPSO 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
H56187-22
H56187-30
AAH56187-22EA
226
USD
AAH56187-22
AAH56187-30
DIPSO 99%
DIPSO
Purity: 99%
CAS No.: 68399-80-4
Molecular Formula: C7H17NO6S
Formula Weight: 261.29
Beilstein Reference: 2105454
MDL No.: MFCD00038353
Formula:
C₇H₁₇NO₆S MW: 261.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038353 CAS Number: 68399-80-4 EINECS: 269-992-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...