2-Formyl-3-picoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylpyridine-2-carboxaldehyde, 3-Methylpicolinaldehyde

H56168-03 H56168-06
AAH56168-03EA 109 USD
AAH56168-03 AAH56168-06
2-Formyl-3-picoline 97%
2-Formyl-3-picoline

Purity: 97% CAS No.: 55589-47-4 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 73°(163°F) Refractive Index: 1.538 MDL No.: MFCD02181144

Formula: C₇H₇NO
MW: 121.14 g/mol
Density: 1.08
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD02181144
CAS Number: 55589-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR