2,4-Dichloro-5-methoxypyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H56153-09
H56153-22
AAH56153-09EA
86.7
USD
AAH56153-09
AAH56153-22
2,4-Dichloro-5-methoxypyrimidine 97%
2,4-Dichloro-5-methoxypyrimidine
Purity: 97%
CAS No.: 19646-07-2
Molecular Formula: C5H4Cl2N2O
Formula Weight: 179.00
Melting Point: 66-70°
MDL No.: MFCD02685670
Formula:
C₅H₄Cl₂N₂O MW: 179.00 g/mol Melting Pt: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD02685670 CAS Number: 19646-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...