3,4-Dihydro-2H-pyran-2-methanol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxymethyl-3,4-dihydro-2H-pyran, DHP Linker
H56124-03
H56124-06
AAH56124-03EA
37.1
USD
AAH56124-03
AAH56124-06
3,4-Dihydro-2H-pyran-2-methanol 96%
3,4-Dihydro-2H-pyran-2-methanol
Purity: 96%
CAS No.: 3749-36-8
Molecular Formula: C6H10O2
Formula Weight: 114.14
Storage Temperature: 2°C to 8°C
Flash Point: 89°(192°F)
Refractive Index: 1.4780
Boiling Point: 92-93°/25mm
MDL No.: MFCD00051507
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 92…93 °C/25 mm Density: 1.101 Flash Pt: 89 °C(192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051507 CAS Number: 3749-36-8 EINECS: 223-150-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...