2-Fluoro-N-methylbenzamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-n-methylbenzamide
H56104-03
H56104-MD
AAH56104-03EA
318
USD
AAH56104-03
AAH56104-MD
2-Fluoro-N-methylbenzamide 97%
2-Fluoro-N-methylbenzamide
Purity: 97%
CAS No.: 52833-63-3
Molecular Formula: C8H8FNO
Formula Weight: 153.16
MDL No.: MFCD01214098
Formula:
C₈H₈FNO MW: 153.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01214098 CAS Number: 52833-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...