3-Boc-3-methylamino azetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H56060-03 H56060-MD
AAH56060-03EA 246 USD
AAH56060-03 AAH56060-MD
3-Boc-3-methylamino azetidine hydrochloride 95%
3-Boc-3-methylamino azetidine hydrochloride

Purity: 95% CAS No.: 577777-20-9 Molecular Formula: C9H18N2O2.HCl Formula Weight: 222.72 MDL No.: MFCD12963843

Formula: C₉H₁₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12963843
CAS Number: 577777-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR